Skip to Content
Merck
All Photos(1)

Key Documents

G006299

LC/MS Analysis of Opioid Glucuronide Metabolites in Urine on Ascentis® Express F5 after Solid Phase Extraction (SPE) using Supel-Select HLB

application for SPE, application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable
solid phase extraction (SPE): suitable

test parameters

sample preparation: Solid Phase Extraction
sample/matrix: urine spiked with drugs and metabolites at 300 ng/mL
SPE well plate: Supel-Select HLB 96-well plate, 60 mg/well (575662-U)
condition: 1 mL methanol followed by 1 mL water
sample addition: 1 mL hydrolyzed urine
washing: 1 mL water followed by 1 mL 25% methanol
elution: 1 mL acetonitrile
column: Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase: [A] 5 mM ammonium formate, pH 4.2 (adjusted with formic acid); [B] 5 mM ammonium formate in (95:5 acetonitrile:water) pH 5.2 (adjusted with formic acid)
gradient: 15% B held for 1 min, to 70% B in 2 min, held at 70% B for 7 min
flow rate: 0.4 mL/min
column temp.: 35 °C
detector: ESI(+) TOF, full scan, m/z 100-1000
injection: 3 μL
sample: 300 ng/mL

suitability

application for HPLC
application for SPE

application(s)

clinical
forensics and toxicology

Analysis Note

Sample pretreatment: β-glucuronidase hydrolysis
The workflow for the LC/MS analysis of native drugs and associated metabolites from urine is shown here. The convenient liquid form of beta-glucuronidase from Sigma was used for hydrolysis of the glucuronide metabolites. SPE using Supel-Select HLB yielded clean extracts. The Ascentis Express F5 columns gave highly efficient, well-resolved peaks and rapid separation. Ultra high purity LC-MS solvents and additives provided trouble-free operation, and Cerilliant CRMs gave reliable quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Supel is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Oxymorphone-3-β-D-glucuronide solution, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®
    Morphine-3-β-D-glucuronide solution, 1 mg/mL in methanol with 0.05% NaOH, ampule of 1 mL, certified reference material, Cerilliant®
    Hydromorphone-3-β-D-glucuronide, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®
    Codeine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®
    Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Oxymorphone-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Hydromorphone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Norbuprenorphine glucuronide solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Codeine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Hydrocodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Oxycodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Buprenorphine glucuronide solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Norbuprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Buprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    See All (14)

SPE tube or plate

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service