Skip to Content
Merck
All Photos(1)

Key Documents

G006131

HPLC Analysis of Yeast Lysate on Proteomix® SAX-NP3 Demonstrating High Separation Efficency

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column temp.: 25 °C
column: Proteomix SAX-NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777219)
detector: UV, 280 nm
flow rate: 0.5 mL/min
gradient: 0 - 100% B in 30 min
injection: 40 μL
mobile phase: (A) 20 mM Tris, pH 9.0; (B) A + 0.5 M NaCl
sample: 2.5 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service