G005688
LC/MS Analysis of Conjugated Bile Acids on Ascentis® Express C18
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Ascentis Express C18, 5 cm x 4.6 mm I.D., 2.7 μm particles (53826-U)
mobile phase: [A] 10 mM ammonium acetate, pH 4 with acetic acid; [B] acetonitrile
gradient: 25 to 40% B in 10 min
flow rate: 1.0 mL/min (postcolumn split to MS)
column temp.: 35 °C
detector: MS, ESI(-), SIR
injection: 5 μL
sample: 10 mg/L each in mobile phase A
suitability
application for HPLC
application(s)
clinical
Legal Information
Analyte
analytical column
mobile phase component
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service