Skip to Content
Merck
All Photos(1)

Key Documents

G005625

HPLC Analysis of Water-Soluble B-Vitamins on Ascentis® Express C18

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express C18, 5 cm x 3.0 mm I.D., 2.7 μm particles (53811-U)
mobile phase: [A] 20 mM potassium phosphate, dibasic, pH 7; [B] methanol
gradient: 0.5% B for 1.3 min, 0.5-30%B over 1.7 min; to 30% B in 1.4 min, 2 min equilibration at 0.5% B
flow rate: 1.0 mL/min
pressure: 4130 psi (285 bar)
column temp.: 35 °C
detector: UV, 210 nm
sample: 9 vitamins in water, 10 μg/mL for nicotinic acid and folic acid, 12 μg/mL for thiamine hydrochloride and cyanocobalamin, (20 μg/mL for pyridoxine, 30 μg/mL for riboflavin, 40 μg/mL for niacineamide, 100 μg/mL for calcium pantothenate, 120 μg/mL for biotin)

suitability

application for HPLC

application(s)

vitamins, nutraceuticals, and natural products

Analysis Note

A mix of 9 water-soluble B-vitamins was separated on Ascentis Express C18 column

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Nicotinic acid, ≥99.5% (HPLC)
    Calcium pantothenate, meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10)
    Pyridoxine hydrochloride, ≥98% (HPLC)
    Nicotinamide, ≥98.5% (HPLC)
    Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
    Biotin, ≥99.0% (T)
    Folic acid, analytical standard
    Cyanocobalamin (B12), analytical standard
    Riboflavin (B2), analytical standard
    See All (9)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service