Skip to Content
Merck
All Photos(1)

Key Documents

FFA57298-U

Supelco

SPME Fiber Assembly

needle size 23 ga, matrix active group 50/30µm DVB/CAR/PDMS, for use with multi fiber exchanger, Stableflex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.21

material

fused silica SPME fiber
plain gray hub

needle size

23 ga

packaging

pkg of 3 ea

df

30 μm (CAR/PDMS layer)
50 μm (DVB layer)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

50/30µm DVB/CAR/PDMS
DVB/CAR/PDMS coating

application(s)

food and beverages

compatibility

for analyte group flavors (volatiles and semivolatiles, C3-20) (MW 40-275)
for use with multi fiber exchanger

Looking for similar products? Visit Product Comparison Guide

Application

SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) may be used for the trapping and extraction of volatile organic compounds in cultures of filamentous fungi[1] and water samples{3 using head-space solid phase microextraction (SPME) coupled to gas chromatography-mass spectrometry (GC-MS).[1][2]

Other Notes

Although we no longer distribute SPME-FFA products andaccessories, these products are available and distributed exclusively byChromline. Please visit the Chromline websiteor contact [email protected] for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Identification and profiling of volatile metabolites of the biocontrol fungus Trichoderma atroviride by HS-SPME-GC-MS.
Stoppacher N, et al.
Journal of Microbiological Methods, 81(2), 189-193 (2010)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)
Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)

Related Content

SPME fast-fit fiber assemblies simplify method development for solid-phase microextraction with easy installation and color-coded labels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service