Skip to Content
Merck
All Photos(1)

Documents

90958

Supelco

2-Hydroxy-4-(octyloxy)benzophenone

analytical standard

Synonym(s):

2-Benzoyl-5-octyloxyphenol, Benzophenone, 2-hydroxy-4-(octyloxy), [2-Hydroxy-4-(octyloxy)phenyl]phenylmethanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
326.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

mp

47-49 °C (lit.)

SMILES string

CCCCCCCCOc1ccc(c(O)c1)C(=O)c2ccccc2

InChI

1S/C21H26O3/c1-2-3-4-5-6-10-15-24-18-13-14-19(20(22)16-18)21(23)17-11-8-7-9-12-17/h7-9,11-14,16,22H,2-6,10,15H2,1H3

InChI key

QUAMTGJKVDWJEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service