854177-U
Push-On Caps for 20 mm headspace vials
polyethylene cap, PTFE-faced butyl septum, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE-faced butyl septum
polyethylene cap
feature
closure type crimp seal (4.3mm opening)
packaging
pkg of 100 ea
fitting
thread for 20 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Seals for washer bottles on the instruments
- Intermediate closure when collecting samples in the field
- Suitable for bevel top headspace vials
These caps can be used for washer bottles with a 20 mm crimp seal. They can also be used as a intermediate closure if a metalic crimp seal is not available.
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service