Skip to Content
Merck
All Photos(1)

Documents

823426

TOYOPEARL® AF-rProtein A HC-650F Bulk Media

phase rProtein A, bottle of 25 mL, 45 μm particle size

Synonym(s):

TOYOPEARL® AF-rProtein A HC-650F Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500

description

Adsorption capacity (> 80 g/L IgG (static); >70 g/L IgG (dynamic at 5 min. residence time))
Phase filter (Protein A)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 23426

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

rProtein A phase

particle size

45 μm

pore size

1000 Å

cleaning in place

0.1 M NaOH

pH

2-13

capacity

>70 g/L adsorption capacity (IgG)(dynamic at 5 min. residence time)
>80 g/L adsorption capacity (IgG)(static)

separation technique

affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

TOYOPEARL AF-rProtein A HC-650F is a high capacity Protein A resin for monoclonal antibody purification. An enhanced rProtein A ligand is bound to the TOYOPEARL HW-65F base bead via multipoint attachment resulting in excellent base stability for up to 200 CIP cycles with 0.1 mol/L NaOH.
TOYOPEARL AF-rProtein A HC-650F resin maintains 80% of initial dynamic binding capacity after 40 CIP cycles with 0.5 mol/L NaOH. TOYOPEARL AF-rProtein A HC-650F resin exhibits dynamic binding capacities of greater than 65 g/L at residence times of 5 minutes and greater than 50 g/L at 2 minutes residence time with feed stock concentrations from 1.0 g/L to 10.0 g/L.
Achievement of high linear velocities at relatively low pressure enables high throughput at production scale using equipment with moderate pressure limitations. Improved mass transfer characteristics allow it to maintain a larger percent of its capacity at lower residence times relative to agarose base stable resins. Typical leakage for this rProtein A ligand is 0.6 -1.7 ng rProtein A /mg eluted antibody by ELISA testing. Though TOYOPEARL AF-rProtein A-650F resin is shipped in 20% ethanol, it can be stored in 2% benzyl alcohol if necessary.
Good pressure-flow characteristics and kinetics contribute to maintaining good capacity particularly at low residence times.

Specifications

Clean in place with 0.1 M NaOH.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service