Skip to Content
Merck
All Photos(1)

Key Documents

807656

TSKgel® Phenyl-5PW (13 µm) HPLC Columns

L x I.D. 15 cm × 21.5 mm, phenyl phase

Synonym(s):

TSKgel® Hydrophobic Interaction Chromatography (HIC) Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£5,970.00

£5,970.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
£5,970.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

£5,970.00


Please contact Customer Service for Availability

Product Name

TSKgel® Phenyl-5PW HPLC Column, phase phenyl, L × I.D. 15 cm × 21.5 mm, 13 μm particle size, stainless steel hardware

material

stainless steel hardware

product line

TSKgel®

manufacturer/tradename

Tosoh Phenyl-5PW, 07656

parameter

≤4 M salt concentration
≤50% organic solvent
10-50 °C temperature
20 bar max. pressure
8 mL/min flow rate

technique(s)

HPLC: suitable

L × I.D.

15 cm × 21.5 mm

matrix

polymer particle platform

matrix active group

phenyl phase

particle size

13 μm

pore size

1000 Å

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity and allow selective binding and desorption of proteins. However, HIC operates at significantly lower binding energy and uses aqueous mobile phases. These characteristics provide a gentle technique that is less likely to disturb protein conformation. As a result, HIC generally provides better activity recovery.

TSKgel BioAssist Phenyl columns contain the same particles as Phenyl-5PW columns, but are packed in PEEK column hardware rather than stainless steel. BioAsist Phenyl (Phenyl-5PW), TSKgel Ether-5PW and Butyl-NPR resin-based columns provide a range of hydrophobicities for chromatographic optimization. Ammonium sulfate concentration can be minimized by using a more hydrophobic packing, such as BioAssist Phenyl. We recommend an Ether-5PW column for purifying very hydrophobic proteins. TSKgel BioAssist Phenyl and TSKgel Ether-5PW packings are based on TSKgel G5000PW resin - 10 μm particles with 1000 Å pores. TSKgel Butyl-NPR columns, which are less hydrophobic than Ether-5PW, is prepared from 2.5 μm nonporous particles, allowing for rapid analyses and semi-micropreparative applications. All three column types can be cleaned with 0.2 M NaOH. Scale-up can be performed by using corresponding Toyopearl bulk resins.
The most hydrophobic among the three TSKgel HIC columns, TSKgel Phenyl-5PW is an excellent choice to screen for the selectivity, retention and recovery of most biomolecules. Featuring a 1000Å pore size in a polymethacrylate base material, the Phenyl-5PW column is stable in either acid or caustic cleaning regimens and provides excellent access to larger molecules with low diffusion coefficients. Stainless steel is the standard column format for TSKgel Phenyl-5PW. Glass and PEEK column housings are available for samples that may adsorb to stainless steel. The alternative column formats are also suitable for general applications.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service