Skip to Content
Merck
All Photos(1)

Key Documents

807164

TSKgel® DEAE-5PW (10 µm) HPLC Columns

L x I.D. 7.5 cm × 7.5 mm, DEAE (diethylaminoethyl) phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£1,770.00

£1,770.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
£1,770.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

£1,770.00


Please contact Customer Service for Availability

Product Name

TSKgel® DEAE-5PW HPLC Column, phase DEAE (diethylaminoethyl), L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size

material

PEEK

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh 07164

parameter

≤1 M salt concentration
≤20% organic solvent
1.2 mL/min flow rate
10-45 °C temperature
150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 7.5 mm

matrix

polymer particle platform

matrix active group

DEAE (diethylaminoethyl) phase

particle size

10 μm

pore size

1000 Å

operating pH

2-12

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel DEAE-5PW columns are packed with porous hydrophilic polymer beads which are surface modified with a weak anion exchanger. They are used for the separation and analysis of proteins, nucleotides, nucleosides and other biologically active molecules. Columns are available in internal diameters varying from 2mm to 55mm ID. Column housings are either glass or stainless steel. They have the same backbone chemistry and selectivity as the bulk process scale resins Toyopearl DEAE-650 and TSKgel DEAE-5PW (20) and (30).

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What should be done before restarting the use of a DEAE column, and which buffer can be used to check if the DEAE column is still working?

    1 answer
    1. If the column does not perform as expected after rehydration and running the QA test method, it should be cleaned according to the instructions. Then, the mobile phase should be equilibrated again, and the QA method should be rerun. If the column still does not perform as expected, it may not be regeneratable and will need to be replaced.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service