69683
2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol
analytical standard
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
80-83 °C (lit.)
format
neat
storage temp.
2-30°C
SMILES string
CCC(C)(C)c1cc(c(O)c(c1)C(C)(C)CC)-n2nc3ccccc3n2
InChI
1S/C22H29N3O/c1-7-21(3,4)15-13-16(22(5,6)8-2)20(26)19(14-15)25-23-17-11-9-10-12-18(17)24-25/h9-14,26H,7-8H2,1-6H3
InChI key
ZMWRRFHBXARRRT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - STOT RE 2 Oral
Target Organs
Liver,Kidney
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service