Skip to Content
Merck
All Photos(1)

Key Documents

63338-U

Supelco

BIOshell lgG 1000 Å C4 (2.7 μm) HPLC Columns

L × I.D. 5 cm × 500 μm, HPLC Column

Synonym(s):

BIOshell IgG 1000 Å C4, 2.7 μm HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
£805.00

£805.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
£805.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

£805.00


Please contact Customer Service for Availability

Product Name

BIOshell lgG 1000 Å C4, 2.7 μm HPLC Column, column L × I.D. 5 cm × 500 μm

material

stainless steel hardware

Quality Level

Agency

suitable for USP L26

description

Shell thickness (0.5 μm)
Solid core (1.7 μm)

product line

BIOshell

feature

endcapped

packaging

pkg of 1 unit

extent of labeling

0.6% carbon loading

parameter

90 °C max. temp.

technique(s)

HPLC: suitable

column L × I.D.

5 cm × 500 μm

surface area

22 m2/g

matrix

spherical silica particle platform

matrix active group

C4 (butyl) phase

particle size

2.7 μm

pore size

1000 Å

operating pH range

2-9

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

BIOshell IgG 1000Å C4 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1000Å) Fused-Core® particle design. The Fused-Core particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5-micron thin porous shell and the small overall particle size of 2.7-microns. The densely bonded, extensively endcapped dimethylbutyl stationary phase of BIOshell IgG 1000Å C4 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds such as proteins.

Legal Information

Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service