Key Documents
58630
Flangeless Fittings Kit (Upchurch)
Select a Size
1 EA
£290.00
£290.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
£290.00
About This Item
UNSPSC Code:
41121815
£290.00
Please contact Customer Service for Availability
Request a Bulk Order
Other Notes
All fittings are 1/4-28 unless otherwise indicated.
Kit Components Only
Product No.
Description
  • Flangeless ferrules, 1/16 in. 10 ea
  • Flangeless ferrules, 1/8 in. 10 ea
  • Flangeless nuts, 1/16 in. 10 ea
  • Flangeless nuts, 1/8 in. 10 ea
  • Unions with fittings, 1/16 in. 2 ea
  • Unions with fittings, 1/8 in. 2 ea
  • Tee with fittings, 1/16 in. 1 ea
  • Tee with fittings, 1/8 in. 1 ea
  • Cross with fittings, 1/16 in. 1 ea
  • Cross with fittings, 1/8 in. 1 ea
  • Unions, metric M6 2 ea
  • Unions, standard 1/4−28 2 ea
  • Luer fittings, female 2 ea
  • Luer fittings, male 2 ea
  • Metric adapters, 1/4−28 to M6 2 ea
See All (15)
Choose from one of the most recent versions:
Certificates of Analysis (COA)