Skip to Content
Merck
All Photos(1)

Key Documents

56704

Shutoff Valve, Low Pressure

natural PEEK, for use with 1/8 in. tubing

Synonym(s):

2-WAY SHUT OFF VALVE, PEEK, W/1/8IN FLANGELESS FITTINGS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

Kel-F rotor
natural PEEK

feature

manual

packaging

pkg of 1 ea

manufacturer/tradename

Upchurch Scientific, Inc P-733

parameter

500 psi max. pressure

thru-hole size

0.040 in.

valve internal volume

10.0 μL , maximum, with valve fully open

fitting

1/4-28 UNF fittings for 1/8 in. O.D. tubing (flangeless fittings included)

compatibility

for use with 1/8 in. tubing

Looking for similar products? Visit Product Comparison Guide

General description

  • Designed for lower pressure applications
  • Biocompatible, all polymer flow path

PEEK or Tefzel valve with 1/4-28 fittings, for pressures up to 1000 psi (70 kg/cm2). Flangeless nuts and ferrules included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service