Skip to Content
Merck
All Photos(1)

Key Documents

48596-U

Supelco

DCMA PCB Mixture

certified reference material, in hexane (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 2 × 5 mL

concentration

in hexane (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    2-Chlorobiphenyl 100 μg/mL
    Decachlorobiphenyl 5 μg/mL
    3,3′-Dichlorobiphenyl 100 μg/mL
    2,2′,3,4′,5,5′,6-Heptachlorobiphenyl 5 μg/mL
    2,2′,3,3′,6,6′-Hexachlorobiphenyl 10 μg/mL
    2,2′,3,3′,4,4′,5,5′,6-Nonachlorobiphenyl 5 μg/mL
    2,2′,3,3′,4,4′,5,5′-Octachlorobiphenyl 5 μg/mL
    2,3′,4,5′,6-Pentachlorobiphenyl 10 μg/mL
    2,2′,4,4′-Tetrachlorobiphenyl 10 μg/mL
    2,4,5-Trichlorobiphenyl 10 μg/mL
    See All (10)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service