Skip to Content
Merck
All Photos(1)

Key Documents

44589

Supelco

Olefins Mix

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

analytical standard

Agency

ASTM® D6729,D6730,D6733

packaging

pkg of 0.1 mL

concentration

(typical values shown)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

General description

This is a 25 component mix of known olefins. It is intended for use in identifying olefins in complex hydrocarbon mixtures.
  • Formulation is weight percent and will vary lot to lot.
  • A detailed data sheet listing components by weight percent, mole percent, and liquid volume percent, retention times and other formulation is provided with the product.
  • Supplied in a crimp-top vial with hole cap and septa.

Application

Olefins Mix has been used as an analytical reference standard for the quantification of the analyte obtained as Fischer-Tropsch (FT) organic phase product using gas chromatography technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    1-Decene 7.348 wt. %
    1-Heptene 7.701 wt. %
    cis-2-Heptene 5.821 wt. %
    trans-2-Heptene 3.77 wt. %
    cis-3-Heptene 5.918 wt. %
    trans-3-Heptene 3.725 wt. %
    1-Hexene 7.753 wt. %
    cis-2-Hexene 3.962 wt. %
    trans-2-Hexene 1.776 wt. %
    Isoprene 2.47 wt. %
    2-Methyl-1-butene 1.466 wt. %
    3-Methyl-1-butene 2.015 wt. %
    2-Methyl-2-pentene 3.402 wt. %
    4-Methyl-1-pentene 3.478 wt. %
    1-Nonene 7.778 wt. %
    cis-2-Nonene 2.776 wt. %
    trans-2-Nonene 1.012 wt. %
    cis-3-Nonene 4.074 wt. %
    trans-3-Nonene 1.88 wt. %
    1-Octene 7.804 wt. %
    cis-2-Octene 3.982 wt. %
    trans-2-Octene 2 wt. %
    1-Pentene 4.214 wt. %
    cis-2-Pentene 2.023 wt. %
    trans-2-Pentene 1.849 wt. %
    See All (25)

analytical column

standard

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Automatic gas chromatographic retention time matching applied to synthetic petroleum (Fischer-Tropsch) products, using HP Chemstation software
Snavely K, et al.
Journal of Chromatography A, 791(1-2), 197-202 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service