Skip to Content
Merck
All Photos(1)

Key Documents

442232-U

Supelco

1,2-Diphenylhydrazine

new

analytical standard, ampule of 100 mg

Synonym(s):

Hydrazobenzene, N,N′-Diphenylhydrazine, N,N′-Bianiline, NSC 3510

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHC6H5
CAS Number:
Molecular Weight:
184.24
Beilstein:
639793
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

123-126 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

N(Nc1ccccc1)c2ccccc2

InChI

1S/C12H12N2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10,13-14H

InChI key

YBQZXXMEJHZYMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Diphenylhydrazine is mainly used in the manufacture of fabrics and medicines.[1] Its other applications include, but not limited to: a precursor of benzidine, an antisludging additive for motor oil, a constituent in photochromic resins as well as in polymers, and syntheses of hydrogen peroxide (H2O2) and several dyestuffs.[2][3]

Application

1,2-Diphenylhydrazine may be used as an analytical standard for the determination of the analyte in landfill leachate, pharmaceutical process stream, and aqueous solutions by high-performance liquid chromatography (HPLC) and gas chromatography (GC) based analytical techniques.[4][5][6]
Reactant involved in:
  • Insertion reactions with organometallic tantalum complexes
  • Reduction reactions catalyzed by titanium(III) trichloride yielding amines
  • Studying the mechanism of hydrazobenzene rearrangement
  • Reaction with N-heterocyclic stable silylene
  • Synthesis of dimanganese amide hydrazide cluster complexes
  • Iron-mediated hydrazine reductions yielding iron arylimide cubanes
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joonas Jernberg et al.
Talanta, 103, 384-391 (2012-12-04)
Nontarget analysis means that a sample is analysed without preselection of the studied analytes. While target analysis attempts to determine whether certain selected compounds are present in the sample, nontarget analysis is performed to explore what unknown compounds can be
Darol E Dodd et al.
International journal of toxicology, 31(6), 564-571 (2012-11-09)
Male F344 rats were exposed to hydrazobenzene (HZB) by dietary feed at concentrations of 0, 5, 20, 80, 200, or 300 ppm for 5 days, 2 weeks, 4 weeks, or 13 weeks duration. End points evaluated included clinical observations, body
Hydrazobenzene.
Report on carcinogens : carcinogen profiles, 10, 139-140 (2004-08-26)
1,2-Diphenylhydrazine
Toxicology Desk Reference: The Toxic Exposure & Medical Monitoring Index (1992)
Public Health Statements: What You Need to Know About Toxic Substances Commonly Found at Superfund Hazardous Waste Sites, Part 1
Public Health Statements (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service