Skip to Content
Merck
All Photos(1)

Key Documents

43256

Supelco

Palmitic acid-1,2,3,4-13C4

analytical standard

Synonym(s):

Hexadecanoic acid-1,2,3,4-13C4

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
£91.90
10 MG
£299.00

£91.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
£91.90
10 MG
£299.00

About This Item

Linear Formula:
CH3(CH2)11(13CH2)313CO2H
CAS Number:
Molecular Weight:
260.39
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

£91.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99%

mp

61-64 °C (lit.)

mass shift

M+4

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC[13CH2][13CH2][13CH2][13C](O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i13+1,14+1,15+1,16+1

InChI key

IPCSVZSSVZVIGE-FIZPRHLTSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

402.8 °F - closed cup

Flash Point(C)

206.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service