42352
N-(2,6-Dimethylphenyl)pyridine-2-carboxamide
reference material
Synonym(s):
Mepivacaine impurity C (PhEur)
About This Item
grade
reference material
Assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
InChI
1S/C14H14N2O/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12/h3-9H,1-2H3,(H,16,17)
InChI key
SHZDEASIMRREIQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
replaced by
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service