Skip to Content
Merck
All Photos(1)

Key Documents

28525-U

Supelco

ATIS Replacement Luer/Hose Barb Adapter

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£36.60

£36.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
£36.60

About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

£36.60


Please contact Customer Service for Availability

Request a Bulk Order

application(s)

air monitoring
environmental
industrial hygiene

General description

The ATIS will accept either 1/4 inch or 6 mm O.D. Thermal Desorption tubes. Included is a luer/hose barb adapter to connect a variety of solvent desorption tubes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service