Documents
22802
Chart Paper for Recorder
Kipp & Zonen XR-9, chart paper L × W 66 ft × 230 mm, chart type, 0-100, pkg of 25 roll
About This Item
UNSPSC Code:
14111500
feature
chart type 0-100
packaging
pkg of 25 roll
manufacturer/tradename
Kipp & Zonen XR-9
WESTRONICS
chart paper L × W
66 ft × 230 mm
compatibility
for use with Kipp & Zonen BD 40/41
General description
The main purpose of a recorder is to record electrical and non-electrical quantity as a function of time. Graphic recorders use elementary rudiments like chart paper for displaying and storing the physical quantity and pen for marking the variation in physical quantity. Chart paper is passed through the recorder at a constant rate. The recorder itself becomes a device for measuring retention time. A certain number of inches or centimetres of chart paper corresponds to a certain number of minutes.
Application
Analog signal from the chromatograph may be connected to a recorder with chart paper operating at a speed of 5 min/in. during an analysis performed to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)