Skip to Content
Merck
All Photos(1)

Documents

22006

Swagelok® Reducer

Swagelok® 200-R-4, brass, 1/8 in. Swagelok, for 1/4 in. tube

Synonym(s):

Swagelok® 200-R-4,BRASS REDUCER 1/8X1/4 PK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717

material

brass

packaging

pkg of 2 ea

manufacturer/tradename

Swagelok® 200-R-4

fitting

for 1/4 in. tube
1/8 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nuts and ferrules included.

Application

Brass Swagelok® reducer maybe used for in gas chromatography system for breath hydrogen analysis.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An inexpensive gas chromatograph for breath hydrogen analysis.
T A Robb et al.
Clinica chimica acta; international journal of clinical chemistry, 134(1-2), 235-241 (1983-10-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service