ZRB2699
Anti-CD33/Siglec-3 Antibody, clone 1D10 ZooMAb® Rabbit Monoclonal
recombinant, expressed in HEK 293 cells
Synonym(s):
CD33, Myeloid cell surface antigen CD33, Sialic acid-binding Ig-like lectin 3, Siglec-3
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
recombinant monoclonal
description
clone 1D10
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 39.83 kDa
observed mol wt ~70 kDa
purified by
using Protein A
species reactivity
human, rat, mouse
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... CD33(945)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in U937 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD33/Siglec-3 in U937 cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CD33/Siglec-3 in lysates from EL4 cells and Rat spleen tissue, and His-tagged recombinant fragment of Human CD33/Siglec-3.
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CD33/Siglec-3 in Human tonsil tissue sections.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD33/Siglec-3 in THP-1 cells.
Flow Cytometry Analysis: 1 µg from a representative lot detected CD33/Siglec-3 in one million Human peripheral blood mononuclear cells (PBMC).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service