ZRB2569
Anti-CSF-1R/CD115 Antibody, clone 1F6 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
CSF-1 receptor, Proto-oncogene c-Fms
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1F6, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 107.98 kDa
observed mol wt ~110 kDa
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
monkey
packaging
antibody small pack of 25
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... CSF1R(1436)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in K-562 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CSF-1R/CD115 in K-562 cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CSF-1R/CD115 in Human tonsil lysates.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CSF-1R/CD115 in K-562 cell lysate.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CSF-1R/CD115 in THP-1 cells.
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CSF-1R/CD115 in Human tonsil tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound CSF-1R/CD115 peptide with a KD of 4.3 x 10-9 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service