Skip to Content
Merck
All Photos(1)

Documents

Z768944

ThermalSeal 2® sealing film

W × L 7.94 mm × 13.97 mm, non-sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.35

material

polyester film

sterility

non-sterile

feature

piercable: no

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific TS2-100

technique(s)

ELISA: suitable
PCR: suitable

L

139.7 mm

W × L

7.94 mm × 13.97 mm

W

79.4 mm

General description

These improved films for classic thermal cycling applications consist of a 50 μm polyester film with a 41 μm layer of acrylic adhesive. Compared to the original ThermalSeal® films ThermalSeal® 2 films offer better adhesion, reduced evaporation, longer end tabs for easier handling, and less tendency to tear during removal from the plate.
Each sealing film measures 79.4 x 139.7 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.1 mm. ThermalSeal 2 sealing films are not pierceable.

Legal Information

ThermalSeal is a registered trademark of Excel Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service