Skip to Content
Merck
All Photos(1)

Key Documents

Z744156

Wilmad® 4 mm Rotor for Bruker® MAS Probe

cap, press fit rotor, no o-ring, Kel-F, pack of 1

Synonym(s):

Solid State NMR, MAS NMR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23152100

Pricing and availability is not currently available.

feature

cap press fit rotor

packaging

pack of 1

manufacturer/tradename

Wilmad WP-601-4181

shipped in

ambient

General description

Manufactured from the highest purity Zirconia, MAS Rotors provide the NMR spectroscopist with the ultimate alternative for analysis of solid samples; the need to solvate is eliminated. They are available for most present solid state NMR spectrometers. MAS Rotors are carefully examined for material irregularities by optical methods before and after the precision machining process. Without over-spinning, each rotor is spin tested to only the highest specified spinning speed. Overspin testing, a common practice with some rotor brands, can actually damage the rotor prior to use. High precision (which becomes a necessity for proper spinning performance) is maintained in the manufacture of the end caps*. Most caps are fitted with O-rings for better sealing; some have axial holes for venting. Advantages of Zirconia Rotors Most homogeneous ceramic material structure. Zirconia Rotor Body has higher strength (1,000 MPa, greater than Si3N4). Highest spinning speed High precision superior finish (0.05 µ mirror gloss). Some caps have a uniquely designed detachable O-ring. Use it when a gas-tight environment is required for the sample.

Application

NMR, MAS NMR

Features and Benefits

  • Homogeneous ceramic material structure.
  • Zirconia Rotor Body strength 1,000 MPa
  • Finish 0.05 µ mirror gloss

Legal Information

Bruker is a registered trademark of Bruker-Physik AG
Wilmad is a registered trademark of SP Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service