Documents
Z744071
General Purpose Sealing Film
polypropylene, sterile
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
material
polypropylene
sterility
sterile
feature
RNase and DNase free
packaging
case of 100 ea
manufacturer/tradename
Excel Scientific GPS-PP-100
parameter
-40-120 °C temp. range
General description
  • Certified DNase- & RNase- free
  • Designed to be used with both polystyrene and polypropylene plates
  • 2.0 mil clear polypropylene film features good clarity and easy handling
  • Specially formulated adhesive provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage
  • Removes easily and cleanly from plate surfaces
  • Minimizes aerosols upon film removal
  • Features two end-tabs with perforations for ease of handling, application and removal