Key Documents
Z688029
CELLine Classic bioreactor flask
CL 1000 (for 1L volumes)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.22
material
polystyrene (with 100,000 D MWCO regenerated cellulose semi-permeable membrane)
description
Equivelent to 16 T-225 standard flasks
sterility
sterile; γ-irradiated
usage
Production of monoclonal antibodies and cellular proteins
packaging
case of 3
manufacturer/tradename
Cole Parmer 2440653
capacity
1000 mL
General description
Ideal for MAb and cellular protein production. Viable in vitro alternative to useing mouse ascites. Bioreactors will utilize innovative membrane technology to separate the cell cultivation area from the media chamber. Cell and cellular-secreted product with a MWCO of 10 kD are retained between two non-cytotoxic membranes. The upper semi-permeable membrane allows continuous nutrient diffusion and waste elimination. The lower membrane provides direct oxygenation and ensures optimum gas exchange.
Other Notes
Capable of yeilding concentrated antibodies up to 1-2 mg/mL. Stackable and may be reused with the same cell line under asceptic conditions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)