Z617741
Greiner CELLSTAR® Shorty pipettes
capacity 5 mL
Synonym(s):
Greiner Shorty pipettes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polystyrene
sterility
sterile
feature
RNase and DNase free
graduations Subdivisions 0.1mL
packaging
pack of 200 ea
manufacturer/tradename
Greiner 606190
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Short-format pipettes made of high-grade polystyreneguarantees maximum clarity. Provided with colour codes accordingto international standards. Expiry date and lot number are printed on eachsingle packaging unit. All pipettes are supplied with a filter for protectionagainst the suction of liquid into the pipetting device. Individually wrappedin paper/plastic, with peel-off function and additional breakthrough function.
Features and Benefits
- Sterile
- Free detectable DNase, human DNA, RNase
- Non cytotoxic and non pyrogenic
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service