Skip to Content
Merck
All Photos(1)

Key Documents

Z615358

VivaFlow 200 flipflow filtration

MWCO 5 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
£528.00

£528.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
£528.00

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

£528.00


Please contact Customer Service for Availability

material

polyethersulfone membrane

description

min recirculation volume < 20 mL

sterility

non-sterile

feature

holdup volume 5.3 mL

packaging

pkg of 1 ea

manufacturer/tradename

Sartorius VF20P1

availability

not available in ROW, USA and Canada

membrane area

200 cm2

working volume

200 μL
500—5000 mL

pore size

5 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

Concentrate 250 mL to under 20 mL in just a few minutes or concentrate one liter 50 times in less than 30 minutes. Alternatively use two Vivaflow 200 in parallel and concentrate 5 liters in under 75 minutes. Near total sample recoveries can be expected with most solutions. Desalting or buffer exchange is just as quick when using the optional recirculation assembly available from Sartorius. The economical standard package comes complete with tubing, pressure control gauge, flow restrictor and high pressure pump tubing. All you need is a peristaltic pump capable of handling 6.4 mm O.D. tubing. Should your pump head require larger tubing, link your own peristaltic tube up to the standard product, using the interconnector provided. Vivaflow 200 is easy to clean and can be used repeatedly.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service