Skip to Content
Merck
All Photos(1)

Documents

Z374431

Sigma-Aldrich

Scienceware® cloning discs

diam. 3.2 mm ( 1/8 in.)

Synonym(s):

cloning disc, cloning ring, cloning rings

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106618
NACRES:
NB.22

sterility

sterile; γ-irradiated

packaging

pack of 100 ea

manufacturer/tradename

Bel-Art F37847-0001

diam.

3.2 mm ( 1/8 in.)

Looking for similar products? Visit Product Comparison Guide

General description

Paper discs transfer cells quickly and easily to a 24 well plate for growth. Discs eliminate leak problems associated with cloning cylinders. Risk of contamination and drying out is reduced because cells are exposed only a fraction of the time it would take to apply a cloning cylinder.
Contains 2 vials of 50 cloning discs.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sangwoo Ham et al.
Scientific reports, 7(1), 525-525 (2017-04-04)
Dysfunctional parkin due to mutations or post-translational modifications contributes to dopaminergic neurodegeneration in Parkinson's disease (PD). Overexpression of parkin provides protection against cellular stresses and prevents dopamine cell loss in several PD animal models. Here we performed an unbiased high-throughput

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service