Skip to Content
Merck
All Photos(1)

Documents

Z372404

Sigma-Aldrich

Vertical electrophoresis unit, fixed height, dual gel

Dual mini

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

grade

electrophoresis grade

W × H

11.3 cm × 10 cm

Looking for similar products? Visit Product Comparison Guide

General description

Accepts 10 × 10 cm precast gels
  • Made from high quality acrylic with platinum electrodes
  • Long lasting silicone sponge gasket on upper reservoir for leak-free operation without grease
  • Safety covers with attached safety-tip power leads
  • Designed for use with unique gel-casting gasket; heavy-duty casting clamps compress gasket between glass plates, sealing without grease
For Slab Gel Electrophoresis:
  • Designed to use as little as 200 mL of buffer, and as little as 5 μg of sample per well
  • Use with gel casting accessories which offer the ability to cast one or two gels in a single assembly
  • Use in standard slab gel electrophoresis or in the second dimension of 2D electrophoresis
For Tube Gel Electrophoresis
  • Only the two fixed-height dual-gel units accept tube gel accessories
  • Tubes in four different inside diameters all have the same outside diameter so they are interchangeable in the tube gel adapter
  • Two-sided operation allows up to 20 tubes to be run at a time

Each unit comes with base unit, upper and lower reservoirs with safety covers and attached power leads, and two gel-mounting spring clamps.
Slab and tube gel accessories are not included; order from the accessories list

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service