Z333425
BRAND® BIO-CERT® filter tips, Tip-Set S, sterile
volume range 2-20 μL
About This Item
material
colorless
colorless tip
sterility
(autoclavable at 121 °C)
sterile
feature
ATP free
DNA free
RNase free
barrier
endotoxin free
filter
packaging
pack of 96 ea (Racked)
manufacturer/tradename
BRAND 712122
maximum volume
20 μL
volume range
2-20 μL
Looking for similar products? Visit Product Comparison Guide
General description
- High-quality PP (free from DiHEMDA and oleamide)
- Weight reduction of the 20 μL tips to reduce weight of plastic waste
- Graduations for a quick volume check
- BIO-CERT: sterile, free of DNA, DNase, RNase, endotoxins and ATP
- Autoclavable at 121 °C / 250 °F (2 bar), acc. DIN EN 285
- Environmentally friendly packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service