Key Documents
Z317330
Eppendorf® racks for microcentrifuge tubes
For 24 × 1.5 ml - 2 ml tubes
About This Item
UNSPSC Code:
41103900
Pricing and availability is not currently available.
manufacturer/tradename
Eppendorf® 0030 123.115
Eppendorf® 022364227
availability
not available in USA and Canada
General description
  • 24-place capacity
  • Unique swivel connector to interlock multiple racks for front-to-back and side-to-side configurations
  • Non-slip stand
  • Numbered positions
  • UV-resistant
  • Autoclavable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)