Skip to Content
Merck
All Photos(1)

Key Documents

V5514

Sigma-Aldrich

Anti-Vasopressin V2 Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 3 μg/mL using human kidney

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... AVPR2(554)
mouse ... Avpr2(12000)
rat ... Avpr2(25108)

Immunogen

Synthetic peptide corresponding to the N-termal extracellular domain of human vasopressin V2 receptor. The immunizing peptide has 67% homology with the rat and mouse gene.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Vasopressin V2 receptor is a G-protein-coupled receptor that binds to the peptide hormone arginine vasopressin produced by posterior pituitary gland. The V2 receptor stimulates the production of cyclic AMP, controls the water reabsorption and salt balance by the kidneys. Antagonists to the V2 receptor have applications in the treatment of congestive heart failure, hypertension, hyponatremia and excessive water retention.

Physical form

Solution in phosphate buffered saline containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Garona et al.
Clinical & experimental metastasis, 33(6), 589-600 (2016-05-06)
[V(4)Q(5)]dDAVP is a novel 2nd generation vasopressin analogue with robust antitumour activity against metastatic breast cancer. We recently reported that, by acting on vasopressin V2r membrane receptor present in tumour cells and microvascular endothelium, [V(4)Q(5)]dDAVP inhibits angiogenesis and metastatic progression
Shuai Mu et al.
Molecules (Basel, Switzerland), 19(2), 2694-2706 (2014-02-26)
Twenty-one non-peptide substituted desloratadine class compounds were synthesized as novel arginine vasopressin receptor antagonists from desloratadine via successive acylation, reduction and acylation reactions. Their structures were characterized by 1H-NMR and HRMS, their biological activity was evaluated by in vitro and
Abeda Jamadar et al.
International journal of molecular sciences, 23(14) (2022-07-28)
Vasopressin type-2 receptor (V2R) is ectopically expressed and plays a pathogenic role in clear cell renal cell carcinoma (ccRCC) tumor cells. Here we examined how V2R signaling within human ccRCC tumor cells (Caki1 cells) stimulates stromal cancer-associated fibroblasts (CAFs). We
Natasha Tatiana Sobol et al.
Experimental and therapeutic medicine, 21(6), 566-566 (2021-04-15)
Osteosarcoma is the most prevalent primary bone malignancy. Due to its high aggressiveness, novel treatment strategies are urgently required to improve survival of patients with osteosarcoma, especially those with advanced disease. Desmopressin (dDAVP) is a widely used blood-saving agent that
[Human recombinant alpha-2b interferon (Introna). Intralesional treatment of therapy-resistant condylomata acuminata].
C S Petersen et al.
Ugeskrift for laeger, 149(24), 1594-1595 (1987-06-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service