Skip to Content
Merck
All Photos(1)

Key Documents

V4265

Sigma-Aldrich

VU 590 dihydrochloride hydrate

≥ (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N4O7·2HCl · xH2O
Molecular Weight:
561.46 (anhydrous basis)
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:

form

solid

storage condition

desiccated

solubility

deionized water: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.Cl.[O-][N+](=O)c1ccc(CN2CCOCCOCCN(CCOCC2)Cc3ccc(cc3)[N+]([O-])=O)cc1

InChI

1S/C24H32N4O7.2ClH.H2O/c29-27(30)23-5-1-21(2-6-23)19-25-9-13-33-14-10-26(12-16-35-18-17-34-15-11-25)20-22-3-7-24(8-4-22)28(31)32;;;/h1-8H,9-20H2;2*1H;1H2

InChI key

UARRNDBWOZCTNV-UHFFFAOYSA-N

Biochem/physiol Actions

VU590 inhibits ROMK at sub-micromolar affinity (294 nM) and Kir7.1
VU590 inhibits ROMK at sub-micromolar affinity (294 nM) and Kir7.1 at low micromolar affinity, but it does not inhibit Kir2.1 or Kir4.1. VU590 appears to act by blocking the ion permeation pathway of the channel.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service