Skip to Content
Merck
All Photos(1)

Key Documents

V0755

Sigma-Aldrich

Violuric acid monohydrate

Synonym(s):

2,4,5,6(1H,3H)-Pyrimidinetetrone 5-oxime, 5-Isonitrosobarbituric acid, Alloxan-5-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3N3O4 · H2O
CAS Number:
Molecular Weight:
175.10
Beilstein:
162342
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

240-250 °C (dec.)

SMILES string

O.O\N=C1/C(=O)NC(=O)NC1=O

InChI

1S/C4H3N3O4.H2O/c8-2-1(7-11)3(9)6-4(10)5-2;/h11H,(H2,5,6,8,9,10);1H2

InChI key

YHAIHNZKUCGXRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service