Skip to Content
Merck
All Photos(1)

Key Documents

U1381

Sigma-Aldrich

Uridine 5′-diphospho-N-acetylglucosamine-[glucosamine-6-3H(N)]

aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H27O17N3P2
CAS Number:
Molecular Weight:
607.35
MDL number:
UNSPSC Code:
12352200

form

aqueous ethanol solution

extent of labeling

5-25 Ci/mmol

shipped in

dry ice

storage temp.

−20°C

Physical form

Ethanol: water (70:30) solution

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service