Key Documents
TMO8000-0140
Nalgene® 180 vacuum tubing
tube I.D. × O.D. 3/8 in. × 5/8 in., tube size 50 ft
Synonym(s):
plastic vacuum tubing
About This Item
UNSPSC Code:
41121800
manufacturer/tradename
Nalge-Nunc 8000-0140
tube I.D. × O.D.
3/8 in. × 5/8 in.
tube size
50 ft
wall size
1/8 in.
General description
Transparent, nontoxic, autoclavable plastic tubing has thick walls for high vacuum.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)