T6824
pPolh-FLAG-1 Transfer Vector
Synonym(s):
ddddk tag, dykddddk tag
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
The pPolh-FLAG-1 vector contains the strong viral polyhedrin (polh) promoter for high-level expression of target genes during the very late phase of infection. The vector also contains a high copy bacterial origin of replication and an ampicillin resistance gene (ampr) for easy propagation in Escherichia coli host cells. Following co-transfection with linear baculovirus DNA into insect cells, allelic replacement between homologous viral sequences (AcNPV ORF 603 and ORF 1629) in the vector and the baculovirus DNA transfers the FLAG-target gene fusion sequence into the viral genome.
Application
pPolh-FLAG-1 is a 5575 bp baculovirus transfer vector used for producing FLAGÒ fusion proteins in insect cells. The vector is designed to be compatible with most baculoviral DNA systems that require the essential gene ORF1629 for complementation of lethal deletions and the recovery of viable recombinant virus. It is also a transfer vector for generation of recombinant Baculovirus and expression of N-terminal FLAG® fusions in insect cells.
Legal Information
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Baculovirus Expression Vectors
Baculovirus Expression Vectors: A Laboratory Manual (1994)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service