Skip to Content
Merck
All Photos(1)

Key Documents

T5830

Sigma-Aldrich

Trap-101 hydrochloride

≥98% (HPLC), powder

Synonym(s):

1-[1-(cyclooctylmethyl)-1,2,3,6-tetrahydro-5-(hydroxymethyl)-4-pyridinyl]-3-ethyl-1,3-dihydro-2Hbenzimidazol-2-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H35N3O2·HCl
CAS Number:
Molecular Weight:
434.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CCN1C(=O)N(C2=C(CO)CN(CC2)CC3CCCCCCC3)c4ccccc14

InChI

1S/C24H35N3O2.ClH/c1-2-26-22-12-8-9-13-23(22)27(24(26)29)21-14-15-25(17-20(21)18-28)16-19-10-6-4-3-5-7-11-19;/h8-9,12-13,19,28H,2-7,10-11,14-18H2,1H3;1H

InChI key

YEVMLMQAMCKVSC-UHFFFAOYSA-N

Biochem/physiol Actions

Trap-101 Hydrochloride is a nociceptin/orphanin FQ peptide (NOP) receptor blocker. Trap-101 acts as an inverse agonist. Trap-101 had been found to be an effective NOP receptor antagonist both in vitro and in vivo, with equivalent potency to J113397.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhui Zhao et al.
Diabetes care, 36(11), 3591-3598 (2013-09-26)
Diabetes confers a very high risk of lower-extremity amputation (LEA); however, few studies have assessed whether blood glucose control can reduce LEA risk among patients with diabetes, especially in practice settings where low-income patients predominate. We performed a prospective cohort
Guoliang Li et al.
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery, 27(7), 847-850 (2013-09-26)
To explore the application value of self-made tibial mechanical axis locator in tibial extra-articular deformity in total knee arthroplasty (TKA) for improving the lower extremity force line. Between January and August 2012, 13 cases (21 knees) of osteoarthritis with tibial
Naoki Ozeki et al.
The British journal of ophthalmology, 97(12), 1538-1542 (2013-09-26)
To evaluate the association between the magnitude of relative afferent pupillary defect (RAPD) and the laterality of visual fields in glaucoma patients using pupillography. Evaluations were made with an RAPDx (Konan Medical USA, Irvine, California, USA), which is designed to
Christian Dollendorf et al.
Beilstein journal of organic chemistry, 9, 1652-1662 (2013-09-26)
A redox-responsive polycationic system was synthesized via copolymerization of N,N-diethylacrylamide (DEAAm) and 2-(dimethylamino)ethyl methacrylate (DMAEMA). N,N'-bis(4-chlorobutanoyl)cystamine was used as disulfide-containing cross-linker to form networks by the quaternization of tertiary amine groups. The insoluble cationic hydrogels become soluble by reduction of
Katsumi Shibata et al.
Journal of nutritional science and vitaminology, 59(4), 343-346 (2013-09-26)
Most Japanese women do not consume the estimated average requirement of vitamin B6 (1.7 mg/d) during pregnancy. Nevertheless, these deficiencies are not reported. We investigated a nutritional biomarker of vitamin B6 in pregnant Japanese women as well as their vitamin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service