Skip to Content
Merck
All Photos(1)

Key Documents

T0764

Sigma-Aldrich

TNPAL-galactocerebroside

Synonym(s):

Trinitrophenylaminolauroylgalactocerebroside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H71N5O14
CAS Number:
Molecular Weight:
870.04
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

functional group

amide
hydroxyl

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\C(O)C(COC1OC(CO)C(O)C(O)C1O)NC(=O)CCCCCCCCCCCNc2c(cc(cc2N(=O)=O)N(=O)=O)N(=O)=O

InChI

1S/C42H71N5O14/c1-2-3-4-5-6-7-8-9-10-12-15-18-21-24-35(49)32(30-60-42-41(53)40(52)39(51)36(29-48)61-42)44-37(50)25-22-19-16-13-11-14-17-20-23-26-43-38-33(46(56)57)27-31(45(54)55)28-34(38)47(58)59/h21,24,27-28,32,35-36,39-43,48-49,51-53H,2-20,22-23,25-26,29-30H2,1H3,(H,44,50)/b24-21+

InChI key

CKSSCTIRJIGIPC-DARPEHSRSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service