Skip to Content
Merck
All Photos(2)

Documents

SRP5016

Sigma-Aldrich

CK1 γ 2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CK1G2, CSNK1G2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

25-35 nmol/min·mg

mol wt

~73 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSNK1G2(1455)

General description

Casein kinase 1 γ 2 (CK1γ2) is a member of the CK1 family of serine/threonine protein kinases which play an important role in diverse cell processes, including DNA replication and repair. It is a ubiquitously expressed cytoplasmic kinase. The gene encoding this protein is localized on human chromosome 19p13.3.

Biochem/physiol Actions

Casein kinase 1 γ 2 (CK1γ2) can interact and phosphorylate the metastatic tumor antigen 1 short form (MTA1s) co-localizing it in the cytoplasm. Phosphorylated MTA1s then sequesters the estrogen receptor-α in the cytoplasm of breast cancer cells. CK1γ2 hyperphosphorylates the ceramide transfer protein CERT leading to a decrease in de novo sphingomyelin synthesis. The reduction in synthesis of sphingomyelin is reversed by the expression of CERT mutants that are not hyperphosphorylated.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MTA family of coregulators in nuclear receptor biology and pathology
Bramanandam M
Nuclear Receptor Signaling (2007)
Sandip K Mishra et al.
Oncogene, 23(25), 4422-4429 (2004-04-13)
Recent studies have shown that metastasis-associated protein-1 short form (MTA1s) - metastatic tumor antigen 1 short form sequesters estrogen receptor-alpha (ER-alpha) in the cytoplasm of breast cancer cells. Using a yeast two-hybrid screening to clone MTA1s-interacting proteins, we identified casein
Role for casein kinase 1 in the phosphorylation of Claspin on critical residues necessary for the activation of Chk1
Zheng Meng
Molecular Biology of the Cell (2011)
Sphingolipid Metabolism and Interorganellar Transport: Localization of Sphingolipid Enzymes and Lipid Transfer Proteins
Toshiyuki Yamaji and Kentaro Hanada
Traffic (2014)
Cloning and Chromosomal Mapping of Human Casein Kinase I ?2 (CSNK1G2)
Aya N
Genomics Data (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service