Skip to Content
Merck
All Photos(2)

Documents

SRP5004

Sigma-Aldrich

AMPK (α2/β2/γ2), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AAKG, AAKG2, AMPK, AMPK2, CMH6, H91620p, MGC61468, PRKAA, PRKAA2, PRKAB2, PRKAG2, WPWS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

255-345 nmol/min·mg

mol wt

~36 kDa (B2)
~65 kDa (G2)
~69 kDa (A2)

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

AMP-activated protein kinase (AMPK) exhibits a key role as a master regulator of cellular energy homeostasis. AMPK exists as a heterotrimeric complex composed of a catalytic α subunit and regulatory β and γ subunits. Binding of AMP to the γ subunit allosterically activates the complex. AMPK is activated in response to stresses that deplete cellular ATP (low glucose, hypoxia and ischemia) and via signaling pathways in response to adiponectin, leptin and CAMKKβ.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara B Kahn et al.
Cell metabolism, 1(1), 15-25 (2005-08-02)
The AMP-activated protein kinase (AMPK) is an evolutionarily conserved sensor of cellular energy status, and recent data demonstrate that it also plays a critical role in systemic energy balance. AMPK integrates nutritional and hormonal signals in peripheral tissues and the
D Grahame Hardie
Journal of cell science, 117(Pt 23), 5479-5487 (2004-10-29)
The AMP-activated protein kinase (AMPK) cascade is a sensor of cellular energy status. Whenever the cellular ATP:ADP ratio falls, owing to a stress that inhibits ATP production or increases ATP consumption, this is amplified by adenylate kinase into a much

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service