Skip to Content
Merck
All Photos(2)

Documents

SML3922

Sigma-Aldrich

A-784168

≥98% (HPLC)

Synonym(s):

1-[3-(Trifluoromethyl)pyridin-2-yl]-N-[4-(trifluoromethylsulfonyl)phenyl]-1,2,3,6-tetrahydropyridine-4-carboxamide, 3′-(Trifluoromethyl)-N-{4-[(trifluoromethyl)thio]phenyl}-3,6-dihydro-2H-1,2′-bipyridine-4-carboxamide, A 784168, A784168

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15F6N3O3S
CAS Number:
Molecular Weight:
479.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Brain-penetrant, orally active, potent and selective transient receptor potential cation channel TRPV1 antagonist with in vivo analgesia efficacy.

A-784168 is a brain-penetrant, orally active, potent and selective transient receptor potential (TRP) cation channel TRPV1 antagonist (IC50 = 25/14/33.7/35.1 nM against TRPV1 activation by 50 nM capsaicin/pH 5.5/3 µM NADA/10 µM anandamide in human TRPV1 tranfectants) with little activity or affinity toward other TRP channels and a panel of 74 neurotransmitter receptors. A-784168 reduces capsaicin-induced acute pain (ED50 = 10 µmol/kg p.o. 1h before capsaicin intraplantar injection) and CFA paw injection-induced chronic inflammatory thermal hyperalgesia (EC50 = 54 nmol) in rats in vivo.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of nerve growth factor-induced mechanical and thermal hypersensitivity in rats
European Journal of Pain, 17(4), 469-479 (2013)
Role of TRPV1 and ASIC3 in formalin-induced secondary allodynia and hyperalgesia
Pharmacological Reports, 66(6), 964-971 (2014)
TRPV1 receptors in the CNS play a key role in broad-spectrum analgesia of TRPV1 antagonists
The Journal of Neuroscience, 26(37), 9385-9393 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service