Skip to Content
Merck
All Photos(2)

Key Documents

SML3881

Sigma-Aldrich

AK-2292

≥98% (HPLC)

Synonym(s):

((2-(((2S)-1-((2S)-2-((3-((5-(2-(2,6-Dioxopiperidin-3-yl)-1-oxoisoindolin-4-yl)pent-4-yn-1-yl)(methyl)amino)-3-oxopropyl)(4-(thiazol-2-yl)phenyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)carbamoyl)benzo[b]thiophen-5-yl)difluoromethyl)phosphonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
£106.00
5 MG
£425.00

£106.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
£106.00
5 MG
£425.00

About This Item

Empirical Formula (Hill Notation):
C52H54F2N7O10PS2
CAS Number:
Molecular Weight:
1070.13
UNSPSC Code:
12352200
NACRES:
NA.77

£106.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cell penetrant, potent and highly selective degrader of STAT5A and STAT5B isoforms.


AK-2292 is a cell penetrant, potent and highly selective PROTAC degrader of STAT5A (signal transducer and activator of transcription 5 A) and STAT5B isoforms. AK-2292 potently induces degradation of STAT5 in normal mouse tissues and human chronic myeloid leukemia (CML) xenografts in mice. It potently inhibits the growth of acute myeloid leukemia (AML) cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A selective small-molecule STAT5 PROTAC degrader capable of achieving tumor regression in vivo
Atsunori Kaneshige
Nature Chemical Biology, 19(6), 703-711 (2023)
Discovery of a Potent and Selective STAT5 PROTAC Degrader with Strong Antitumor Activity In Vivo in Acute Myeloid Leukemia
Journal of medicinal chemistry, 66(4), 2717-2743 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service