Skip to Content
Merck
All Photos(1)

Key Documents

SML3863

Sigma-Aldrich

Tecovirimat

≥98% (HPLC)

Synonym(s):

4-Trifluoromethyl-N-(3,3a,4,4a,5,5a,6,6a-octahydro-1,3-dioxo-4,6-ethenocycloprop[f]isoindol-2(1H)-yl)-benzamide, SIGA-246, ST-246, rel-N-[(3aR,4R,4aR,5aS,6S,6aS)-3,3a,4,4a,5,5a,6,6a-Octahydro-1,3-dioxo-4,6-ethenocycloprop[f]isoindol-2(1H)-yl]-4-(trifluoromethyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15F3N2O3
CAS Number:
Molecular Weight:
376.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Antiviral with activity against orthopoxviruses such as cowpox and monkeypox and including smallpox
Tecovirimat (ST-246) is an antiviral with activity against orthopoxviruses such as cowpox and monkeypox and including smallpox. Tecovirimat inhibits the function of a major envelope protein required for the production of extracellar virus, prevented the virus from leaving the cell, and spreading within the body. FDA-approved for the treatment of human smallpox disease caused by Variola virus in adults and children.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficacy of tecovirimat (ST-246) in nonhuman primates infected with variola virus (Smallpox)
Antimicrobial Agents and Chemotherapy, 57(12), 6246-6253 (2013)
Emergence of cowpox: study of the virulence of clinical strains and evaluation of antivirals
PLoS ONE, 8(2), e55808-e55808 (2013)
Fragment-Based Approaches Identified Tecovirimat-Competitive Novel Drug Candidate for Targeting the F13 Protein of the Monkeypox Virus
Viruses, 15(2), 570-570 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service