Skip to Content
Merck
All Photos(2)

Key Documents

SML3727

Sigma-Aldrich

WIC1

≥98% (HPLC)

Synonym(s):

N-[4-(4-Ethyl-1-piperazinyl)phenyl]-2-oxo-2H-1-benzopyran-3-carboxamide, Wnt Inhibitor Compound 1

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
£75.00
50 MG
£303.00

£75.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
£75.00
50 MG
£303.00

About This Item

Empirical Formula (Hill Notation):
C22H23N3O3
CAS Number:
Molecular Weight:
377.44
UNSPSC Code:
12352200
NACRES:
NA.77

£75.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to brown

solubility

DMSO:methanol (1:1): 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C1OC2=C(C=C1C(NC3=CC=C(N4CCN(CC4)CC)C=C3)=O)C=CC=C2

InChI key

YQXKWSNLAVFZJL-UHFFFAOYSA-N

Biochem/physiol Actions

Potent Wnt/β-catenin signaling inhibitor that reduces airway basal stem cell (ABSC) proliferation and induces ciliated cell differentiation.
WIC1 is a potent Wnt/β-catenin signaling inhibitor (IC50 = 10 nM against 24h 5 μM CHIR-induced BEAS2B TCF/LEF reporter activity) that reduces airway basal stem cell (ABSC) proliferation (92% reduction of 4d 2 μM CHIR-induced hABSC proliferation by 1 μM WIC1) and induces ciliated cell differentiation (by 1.9-fold on d14 with 100 nM WIC1; mABSCs under air-liquid interface (ALI) condition) by suppressing β-catenin Y489 phosphorylation (complete inhibition of 24h 5 μM CHIR-induced pY489 in BEAS2B by 1 μM WIC1) and TP63 expression (80% reduction of 9d 1 μM CHIR-induced TP63 in mABSC by 1 μM WIC1; 40% reduction of 48h 5 μM CHIR-induced TP63 in BEAS2B by 1 μM WIC1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cody J Aros et al.
Cell reports, 30(7), 2055-2064 (2020-02-23)
Mechanisms underpinning airway epithelial homeostatic maintenance and ways to prevent its dysregulation remain elusive. Herein, we identify that β-catenin phosphorylated at Y489 (p-β-cateninY489) emerges during human squamous lung cancer progression. This led us to develop a model of airway basal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service