Skip to Content
Merck
All Photos(1)

Key Documents

SML3711

Sigma-Aldrich

DENSPM tetrahydrochloride

≥98% (HPLC)

Synonym(s):

BE 333 HCl, BENSpm HCl, DENSpm 4HCl, N1,N11-Diethylnorspermine tetrahydrochloride, N1,N11-bis(ethyl)norspermine tetrahydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£110.00
25 MG
£440.00

£110.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£110.00
25 MG
£440.00

About This Item

Empirical Formula (Hill Notation):
C13H32N4· 4HCl
CAS Number:
Molecular Weight:
390.26
MDL number:
UNSPSC Code:
41121800
UNSPSC Code:
12352200
NACRES:
NA.21

£110.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

DENSPM HCl is a nontoxic polyamine analog that enhances polyamine catabolism and production of hazardous hydrogen peroxide and N-acetyl-3-aminopropanal by inducing spermidine/spermine-N1-acetyl transferase (SSAT) expression. DENSPM HCl improves stem cell maintenance through reduced translation, exhibits slower proliferation rates in cancer cells, and induces autophagy. It shows greater antitumor activity and less associated toxicity in a human melanoma xenograft model.
Polyamine pathway inhibitor; potent inducer of spermidine/spermine N1-acetyltransferase

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service