Skip to Content
Merck
All Photos(1)

Documents

SML3588

Sigma-Aldrich

J147

≥98% (HPLC)

Synonym(s):

(E)-N-(2,4-dimethylphenyl)-2,2,2-trifluoro-N′-(3-methoxybenzylidene)acetohydrazide, 2,2,2-Trifluoro-acetic acid 1-(2,4-dimethylphenyl)-2-[(3-methoxyphenyl)methylene]hydrazide, J 147

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17F3N2O2
CAS Number:
Molecular Weight:
350.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

COC1=CC(/C=N/N(C2=C(C=C(C=C2)C)C)C(C(F)(F)F)=O)=CC=C1

Biochem/physiol Actions

J147 is a broadly neuroprotective (in multiple assays) compound that enhances the memory in normal and Alzheimer’s disease (AD) transgenic mice. J147 has neurotrophic and BDNF-like activities that are independent of the BDNF receptor, and TrkB. Also it appears that J147 has a small but significant effect on Ab metabolism by reducing Ab1–42 in the soluble fraction of the hippocampus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel neurotrophic drug for cognitive enhancement and Alzheimer's disease
Chen Q
PLoS ONE, 6(12) (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service